missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pIB/V5-His TOPO™ TA Expression Kit
Offers five-minute cloning of Taq-amplified PCR products directly into the pIB/V5-His-TOPO expression vector
Brand: Invitrogen™ K89020
759.90 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Includes: Each pIB/V5-His TOPO TA Expression Kit contains two boxes. The pIB/V5-His TOPO box contains 200ng of linearized and topoisomerase I-activated pIB/V5-His-TOPO vector, dNTPs, 10X PCR buffer, control PCR template and primers, forward and reverse primers for sequencing and PCR screening, salt solution, and pIB/V5-His/CAT. The box contains high-efficiency, single-use, 50μL aliquots of chemically competent TOP10 E. coli cells, S.O.C. medium, and pUC19 supercoiled plasmid control.
Description
- The OpIE2 promoter for constitutive expression
- Includes Blasticidin resistance gene for rapid selection of stably transfected cell lines in two weeks
- C-terminal V5 epitope and polyhistidine (6xHis) sequence for detection with Anti-V5 Antibody and rapid purification using nickel-chelating resin
Constitutive Protein Expression in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
OplE2 | |
Each pIB/V5-His TOPO™ TA Expression Kit contains two boxes. The pIB/V5-His TOPO™ box contains 200 ng of linearized and topoisomerase I-activated pIB/V5-His-TOPO™ vector, dNTPs, 10X PCR buffer, control PCR template and primers, forward and reverse primers for sequencing and PCR screening, salt solution, and pIB/V5-His/CAT. Store at -20°C. The One Shot™ box contains high-efficiency, single-use, 50 μl aliquots of chemically competent TOP10 E. coli cells, S.O.C. medium, and pUC19 supercoiled plasmid control. Store at -80°C. All reagents are guaranteed stable for 6 months when properly stored. | |
His Tag (6x), V5 Epitope Tag | |
20 reactions | |
InsectSelect™, TOPO™ |
TOPO TA Expression Kit | |
C-terminal | |
TOPO-TA | |
pIB, TOPO-TA Cloning Vectors |
For Research Use Only. Not for use in diagnostic procedures.