Learn More
Invitrogen™ pIZ/V5-His Vector Kit
Product Code. 10674383
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
For Research Use Only. All usage must comply with product instructions.
Product Code. 10674383
Brand: Invitrogen™ V800001
For Research Use Only. All usage must comply with product instructions.
Includes
The InsectSelect Kit includes 20μg each of supercoiled, lyophilized pIZ/V5-His and pIZ/V5-His/CAT, 2μg of lyophilized OpIE2 Reverse Sequencing primer, 1g Zeocin, Cellfectin Reagent, and 1L of medium for the insect cell line chosen. Sf9 cells are supplied with GIBCO Grace's Insect Media, High Five Cells are supplied with GIBCO Express Five Serum-Free Medium. The pIZ/V5-His Vector Kit contains only the expression vectors and primer as described above.
For high-level expression in a variety of insect cells
  • This exceptionally small vector has several features that facilitate expression, analysis, and detection of recombinant protein in insect cells
  • The OpIE2 promoter for constitutive expression
  • The Zeocin resistance gene for rapid selection of stably transfected cell lines
  • C-terminal V5 epitope and polyhistidine (6xHis) sequence for detection with Invitrogen's Anti-V5 Antibody and rapid purification using nickel-chelating resin
Constitutive Protein Expression in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter OplE2
Product Type Insect Cell Expression Vector
Content And Storage The InsectSelect™ Kit includes 20 μg each of supercoiled, lyophilized pIZ/V5-His and pIZ/V5-His/CAT, 2 μg of lyophilized OpIE2 Reverse Sequencing primer, 1 g Zeocin™, Cellfectin™ Reagent, and 1 L of medium for the insect cell line chosen. Sf9 cells are supplied with GIBCO™ Grace's Insect Media, High Five™ Cells are supplied with GIBCO™ Express Five™ Serum-Free Medium. The pIZ/V5-His Vector Kit contains only the expression vectors and primer as described above. Store frozen insect cells in liquid nitrogen. Store medium and Cellfectin™ Reagent at +4°C. Store all other reagents at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Protein Tag Position (to your gene) C-terminal
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 1 kit
Vector pIZ
Product Line InsectSelect