Learn More
Invitrogen™ Plasmid pBR322
Prepacked column
Brand: Invitrogen™ 15367014
177.08 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: pBR322 is supplied at 0.25μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA.
Description
Plasmid pBR322 (1) contains three unique restriction endonuclease recognition sites within the !-lactamase (AmpR) gene and eight unique sites within the TetR gene. The plasmid, purified from DH10B E. coli, also has 14 non-selectable unique restriction endonuclease recognition sites. Unique cleavage sites for Hind III and Cla I are found in the promoter of the tetracycline resistance gene. Insertion of DNA at either of these two sites usually results in loss of tetracycline resistance. Performance and Quality Testing: Purity, DNA structure, and selected restriction endonuclease fragmentation patterns are verified by agarose gel analysis. Transformation of E. coli HB101 to confirm AmpR and TetR is confirmed by plating on selective medium.
Cloning, Restriction Enzyme Cloning
Order Info
Approved for shipment on Wet or Dry ice
Specifications
Restriction Enzyme/ MCS | |
pBR322 is supplied at 0.25 μg/μl in 10 mM Tris-HCl (pH 7.4), 5 mM NaCl, 0.1 mM EDTA. Store at +4°C. | |
Plasmid | |
Approved for shipment on Wet or Dry Ice |
0.25 μg/μl | |
Cloning | |
25 μg | |
pBR322 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.