missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase

Product Code. p-4919386
Click to view available options
No. of Reactions:
120 Reactions
300 Reactions
5000 Reactions
600 Reactions
Unit Size:
120 reactions
300 reactions
5000 reactions
600 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10033232

Brand: Invitrogen™ 10966018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Each box contains: Platinum Taq DNA Polymerase; 10X PCR Buffer (no magnesium chloride); 50mM magnesium chloride; KB Extender

Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable 'hot start' thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase.

Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable hot-start thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase. The hot start property of the enzyme is conferred by thermolabile monoclonal antibodies that render Taq DNA polymerase inactive until the initial PCR denaturation step, thus preventing the extention of nonspecifically annealed primers and improving product yield.

The hot-start property allows for convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity. PCR products generated with Platinum Taq DNA Polymerase may be used in the same downstream applications without protocol modifications.

Features

  • Convenient reaction assembly at room temperature
  • 5' to 3' exonuclease activity
  • Lacks 3' to 5' exonuclease activity

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • Hot-start PCR
  • RT-PCR

Notes

  • For superior PCR performance, the next-generation enzyme Platinum II Taq Hot-start DNA Polymerase is recommended.
  • Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.

Order Info

Shipping Condition: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Platinum Taq DNA Polymerase, 24 μL
• 10X PCR Buffer (without magnesium), 1.25 mL
• 50 mM MgCl2, 1 mL
• KB Extender, 1.25 mL

Store at -10°C to -30°C in a non-frost-free freezer.
Format Tube
GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Fast or Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Quantity 120 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 120 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.