missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase, High Fidelity

Product Code. 10675783
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
5000 Reactions
Unit Size:
100 reactions
500 reactions
5000 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10675783

Brand: Invitrogen™ 11304102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Platinum Taq DNA Polymerase High Fidelity (1000μL, at 5U/μL) is supplied with a bottle (50mL) of 10X High Fidelity Buffer (600mM Tris-SO4 (pH 8.9), 180mM (NH4)2SO4), and a bottle (25mL) of 50mM MgSO4.

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required.

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum Taq DNA Polymerase and the proofreading enzyme. PCR specificity is improved with built-in Platinum hot-start technology.

Features of Platinum Taq DNA Polymerase, High Fidelity

  • Greater than six times higher fidelity than Taq DNA polymerase
  • Amplification of fragments up to 15 kb
  • Convenient room temperature reaction assembly
  • Increased specificity, and yield

Applications

  • Amplification of complex DNA templates
  • Amplification of viral and plasmid templates
  • RT-PCR

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Platinum Taq DNA Polymerase High Fidelity (1 x 1000 μL at 5 U/μL)
• 10X High Fidelity Buffer (1 x 50 mL)
• 50 mM MgSO4 (1 x 25 mL)

Store at -10°C to -30°C.
Detection Method Primer-probe
GC-Rich PCR Performance Low
PCR Method 1-step RT-qPCR
Polymerase Platinum Taq DNA Polymerase High Fidelity
Reaction Speed Standard
Product Type DNA Polymerase
Quantity 5,000 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 6X
Hot Start Built-In Hot Start
No. of Reactions 5000 Reactions
Overhang Mixed, 3'-A
Reaction Format Separate Components
Size (Final Product) 20 kb or less
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.