missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG

Product Code. 10543283
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10543283

Brand: Invitrogen™ 11730025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

2X concentration [40mM Tris-HCl (pH 8.4), 100mM KCl, 6mM MgCl2 , 400μM dATP, 400μM dCTP, 400μM dGTP, 800μM dUTP, 40 units/mL UDG, 60 units/ml Platinum™ Taq DNA Polymerase, stabilizers], vial of 50mM MgCl2

Is a ready-to-use mixture for high-specificity, real-time quantitative DNA amplification, containing UDG to prevent carryover contamination (1,2).

  • Highly sensitive and specific
  • Allows room temperature reaction assembly
  • Easy to use: just add template and fluorogenic primers or probes for detection
  • Includes a vial of 50mM MgCl2 to accommodate a variety of primer/template PCR systems
  • Compatible with most real-time instruments

Order Info

Guaranteed stable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days. Shipping Conditions: Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Platinum™ Quantitative PCR SuperMix-UDG is supplied in a 2X concentration [40 mM Tris-HCl (pH 8.4), 100 mM KCl, 6 mM MgCl2 , 400 μM dATP, 400 μM dCTP, 400 μM dGTP, 800 μM dUTP, 40 units/ml UDG, 60 units/ml Platinum™ Taq DNA Polymerase, stabilizers]. A vial of 50 mM MgCl2 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored.
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
Product Line Platinum
Product Type Quantitative PCR SuperMix-UDG
Quantity 500 reactions
Shipping Condition Dry Ice
Sufficient For 500 Reactions
For Use With (Application) Gene Expression
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.