missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
Beskrivelse
Indeholder
2X concentration [40mM Tris-HCl (pH 8.4), 100mM KCl, 6mM MgCl2 , 400μM dATP, 400μM dCTP, 400μM dGTP, 800μM dUTP, 40 units/mL UDG, 60 units/ml Platinum™ Taq DNA Polymerase, stabilizers], vial of 50mM MgCl2
- Highly sensitive and specific
- Allows room temperature reaction assembly
- Easy to use: just add template and fluorogenic primers or probes for detection
- Includes a vial of 50mM MgCl2 to accommodate a variety of primer/template PCR systems
- Compatible with most real-time instruments
Ordreinfo
Guaranteed stable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days. Shipping Conditions: Dry Ice.
Tekniske data
Tekniske data
| Content And Storage | Platinum™ Quantitative PCR SuperMix-UDG is supplied in a 2X concentration [40 mM Tris-HCl (pH 8.4), 100 mM KCl, 6 mM MgCl2 , 400 μM dATP, 400 μM dCTP, 400 μM dGTP, 800 μM dUTP, 40 units/ml UDG, 60 units/ml Platinum™ Taq DNA Polymerase, stabilizers]. A vial of 50 mM MgCl2 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ |
| Product Line | Platinum |
| Product Type | Quantitative PCR SuperMix-UDG |
| Quantity | 100 reactions |
| Shipping Condition | Dry Ice |
| Vis mere |
For Research Use Only. Not for use in diagnostic procedures.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion