Learn More
Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit
Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.
£494.00 - £2075.00
Specifications
Detection Method | Primer-probe |
---|---|
PCR Method | 1-step RT-qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ |
Includes: 100μL SuperScript III/Platinum Taq Mix; 2 x 1.25mL 2X Reaction Mix; 1mL Magnesium Sulfate (50mM); 100μL ROX Reference Dye (25μM)
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10442882
|
Invitrogen™
11732020 |
100 Reactions |
£494.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10104912
|
Invitrogen™
11732088 |
500 Reactions |
£2075.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection.
- The SuperScript III Platinum One-Step qRT-PCR Kit’s proven formula delivers reliable, accurate target detection when it matters most
- A convenient, one-step format and consistent performance make it ideal for a number of gene expression applications, including infectious disease surveillance
- The kit has been validated by several global public health organizations and has long been the choice of the US CDC for infectious disease surveillance
- SuperScript III RT for higher temperature cDNA synthesis when addressing difficult RNA secondary structure
- Platinum Taq DNA Polymerase with hot-start technology for improved specificity
- LUX fluorogenic primers and dual-labeled fluorogenic probes for effective detection performance
- Validated performance on multiple real-time instrument platforms, including rotor-based systems
- Multiple SuperScript III Platinum One-Step qRT-PCR Kit configurations are available
- Store components at -20°C, ROX Reference Dye must be stored in the dark
Order Info
Shipping Condition: Dry ice
Specifications
Primer-probe | |
Taq DNA Polymerase | |
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ | |
One-Step qRT-PCR Kit | |
Gene Expression, Pathogen Detection, Virus Detection |
1-step RT-qPCR | |
Standard | |
Platinum™, SuperScript™ | |
Dry Ice | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.