Learn More
SuperScript™, Platinum™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit w/ROX
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.
£490.00 - £2040.00
Specifications
Detection Method | Primer-probe |
---|---|
PCR Method | 1-step RT-qPCR |
Polymerase | Platinum Taq DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT System |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11578656
|
SuperScript™, Platinum™
11745500 |
500 Reactions |
£2040.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11568656
|
SuperScript™, Platinum™
11745100 |
100 Reactions |
£490.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. One-Step qRT-PCR kits with ROX include SuperScript™ III Reverse Transcriptase which produces higher yields of full-length cDNA for broader gene representation, Platinum™ Taq DNA Polymerase which activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification and ROX reference dye premixed at optimal concentration for ABI instruments
Two kit configurations allow your choice of detection methods:- SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection
- SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection for use with LUX™ fluorogenic primers or dual-labeled probes (e.g. TaqMan™)
Order Info
Dry Ice
Specifications
Primer-probe | |
Platinum Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System | |
Dry Ice | |
RNA |
1-step RT-qPCR | |
Standard | |
Platinum™, SuperScript™ | |
Gene Expression, Pathogen Detection, Virus Detection |
For Research Use Only. Not for use in diagnostic procedures.