missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum™, SYBR™ Platinum™ SYBR™ Green qPCR SuperMix-UDG
Provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye.
£267.00 - £1158.00
Specifications
Detection Method | SYBR |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Includes: Platinum SYBR Green qPCR SuperMix-UDG (2 × 1.25mL), 50mM Magnesium Chloride (1mL), ROX Reference Dye (100μL), 20X BSA (300μL)
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10633863
|
Platinum™, SYBR™
11733046 |
500 Reactions |
£1158.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10623863
|
Platinum™, SYBR™
11733038 |
100 Reactions |
£267.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results.
- Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target
- 100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
- Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments
- SYBR™ Green I dye for simple and easy detection of real-time product
Order Info
Shipping Conditions: Dry Ice
Specifications
SYBR | |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, GeneAmp 5700, StepOne™, StepOnePlus™, ViiA™ 7 System, Cepheid SmartCycler, BioRad iCycler iQ | |
Real Time PCR SYBR Master Mix | |
Gene Expression |
Tube | |
qPCR | |
Standard | |
Platinum™, SYBR™ | |
Dry Ice | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title