Learn More
Platinum™, SYBR™ GreenER™ SYBR GreenER™ qPCR SuperMix for iCycler™ Instrument
Incorporate the latest high-performance technology to provide the most reliable gene expression data from your iCycler iQ™ or MyiQ™ instrument.
Brand: Platinum™, SYBR™ GreenER™ 1176102K
Includes: Hot-start Taq DNA polymerase, SYBR GreenER fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers
Description
The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.
Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.
- The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
- A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
- An optimized buffer system - improves sensitivity and provides excellent long-term stability
- Uracil DNA glycosylase - reduces carryover contamination in qPCR
Order Info
Shipping Conditions: Dry Ice
Specifications
2X SYBR™ GreenER™ qPCR SuperMix for iCycler™ instrument contains: hot-start Taq DNA polymerase, SYBR™ GreenER™ fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers. Store at 4°C upon receipt. Guaranteed stable for 6 months when properly stored. |
|
High | |
Taq DNA Polymerase | |
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
2000 Reactions |
SYBR | |
qPCR | |
Standard | |
Platinum™, SYBR™ GreenER™ | |
2000 Reactions | |
2X | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.