Learn More
SuperScript™, Platinum™, SYBR™ SuperScript™ III Platinum™ SYBR™ Green One-Step qPCR Kit w/ROX
Combine a powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.
Brand: SuperScript™, Platinum™, SYBR™ 11746500
Description
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. SuperScript™ III Reverse Transcriptase produces higher yields of full-length cDNA for broader gene representation. Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. ROX reference dye premixed at optimal concentration for ABI instruments.
- Provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection
Order Info
Shipping Conditions: Dry Ice
Specifications
Store in freezer (-5 to -30°C). | |
1-step RT-qPCR | |
Standard | |
Platinum™, SYBR™, SuperScript™ | |
Dry Ice | |
Gene Expression | |
500 Reactions |
SYBR | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System | |
Real Time PCR SYBR Master Mix | |
500 Reactions | |
2X | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.