missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SuperScript™, Platinum™, SYBR™ SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit
Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.
Brand: SuperScript™, Platinum™, SYBR™ 11736059
995.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Includes: 100μL SuperScript III RT/Platinum Taq Mix; 2 x 1.25mL 2X SYBR Green Reaction Mix; 1mL Magnesium Sulfate (50mM); 100μL ROX Reference Dye (25μM); 300μL 20X Bovine Serum Albumin (1mg/mL)
Description
The SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection.
- Kit combines the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format
- The SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection
- SuperScript™ III RT for higher temperature cDNA synthesis for greater success with difficult RNA secondary structure
- Platinum™ Taq DNA Polymerase with hot-start technology for improved specificity
- LUX™ Fluorogenic Primers or dual-labeled fluorogenic probes for superior detection performance
- SYBR™ Green I dye for easy and convenient detection
- Validated performance on multiple real-time instrument platforms, including rotor-based systems
- Store all components at -20°C (-80°C for long-term storage)
- ROX Reference Dye must be stored in the dark
Order Info
Shipping Conditions: Dry Ice
Specifications
• 500 μL SuperScript™ III RT/Platinum™ Taq Mix • 12.5 mL 2X SYBR™ Green Reaction Mix • 2 × 1 mL Magnesium Sulfate (50 mM) • 500 μL ROX Reference Dye (25 μM) • 1.3 mL 20X Bovine Serum Albumin (1 mg/mL) Store all components at -20°C (-80°C for long-term storage). ROX Reference Dye must be stored in the dark. |
|
1-step RT-qPCR | |
Standard | |
Platinum™, SYBR™, SuperScript™ | |
Dry Ice | |
Real Time PCR (qPCR) | |
RNA |
SYBR | |
Taq DNA Polymerase | |
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ | |
One-Step qRT-PCR Kit | |
500 Reactions | |
500 Reactions |
For Research Use Only. Not for use in diagnostic procedures.