Learn More
Invitrogen™ Proteinase K, recombinant
Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.
£145.00 - £900.00
Specifications
Concentration | >40 U/mg |
---|---|
Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
Product Type | Proteinase K |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10124532
|
Invitrogen™
25530031 |
1 g |
£900.00
1g |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11578916
|
Invitrogen™
25530015 |
100 mg |
£145.00
100mg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.
- Over a wide pH range–optimal activity between 6.5 and 9.5
- Under denaturing conditions–e.g., in the presence of SDS or urea
- In the presence of metal chelating agents–e.g., EDTA
- At comparatively high temperatures–optimum digestion temperature is 65°C
- Protein Filter: P
Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.
Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.
Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.
Order Info
Approved for shipment at Room Temperature or on Wet Ice
Specifications
>40 U/mg | |
Approved for shipment at Room Temperature or on Wet Ice | |
Proteinase K | |
Powder |
For Research Use Only. Not for use in diagnostic procedures.