Learn More
Invitrogen™ PureLink™ Genomic Plant DNA Purification Kit
The PureLink Genomic Plant DNA Purification Kit allows total DNA purification from a variety of plant samples in under one hour.
Brand: Invitrogen™ K183001
140.67 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
197.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The PureLink Genomic Plant DNA Purification Kit allows total DNA purification from a variety of plant samples such as spinach, tomato, soy, wheat, Arabidopsis, mushroom, alfalfa sprouts, sunflower sprouts, corn husks, soybeans, mushroom, tomato leaves, wheat grass, and Arabidopsis thaliana leaves in under one hour.
- Efficient—Yields high quality DNA with minimal RNA contamination
- Simple DNA Purification from Plant Samples
- PureLink Genomic Plant DNA Purification Kit is suitable for isolating DNA from chloroplasts and variety of plant tissues
- Entire protocol relies on centrifugation of spin columns in absence of organic solvents
- PureLink Genomic Plant DNA Purification Kit is designed to purify DNA from up to 100 mg of plant tissue
Maximize Yields of Total DNA
- High yield of DNA and minimal DNA degradation is achieved by using young plant samples such as leaves and by freezing sample in liquid nitrogen immediately after collection
- Total DNA is eluted in two aliquots of 100μL each to obtain greater DNA yield, with first elution generating between 65-80% DNA recovery and the second elution > 90%
- Typically, DNA isolated using the PureLink Genomic Plant DNA Purification Kit has an A260/A280 > 1.80 when samples are diluted in Tris-HCl (pH 7.5), indicating that the DNA is reasonably free of proteins that could interfere with downstream applications
Isolated DNA can be used for a Number of Downstream Applications:
- Total DNA purification from different plant tissue with the PureLink Genomic Plant DNA Purification Kit is suitable for a variety of applications, including PCR, restriction enzyme digestion, and southern blotting

Specifications
Spin Column | |
Silica Spin Column | |
Plants | |
gDNA | |
Not High-throughput Compatible (Manual) | |
Room Temperature | |
Up to 14.6 μg |
• 12.5 mL Resuspension Buffer R2; room temperature • 5 mL Precipitation Buffer N2; room temperature • 7.5 mL Binding Buffer B4; room temperature • 25 mL Wash Buffer W4; room temperature • 10 mL Wash Buffer W5; room temperature • 10 mL Elution Buffer; room temperature • 0.75 mL 20% SDS; room temperature • 0.75 mL RNase A; room temperature • 50 PureLink Spin Columns with Collection Tubes; room temperature • 50 Collection Tubes; room temperature |
|
Mini | |
100 to 200 μL | |
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization | |
50 Preps | |
40 min. | |
100 mg |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.