missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure Plasmid Miniprep Kit

Product Code. p-4926488
Click to view available options
No. of Reactions:
100 Preps
25 Preps
Unit Size:
100 reactions
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10013552

Brand: Invitrogen™ K210002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The PureLink HiPure Plasmid Miniprep Kit allows isolation of high yields of highly pure plasmid DNA from E.coli in less than 2 hours, without use of any organic solvents or cesium chloride, and contains Low endotoxin levels.

The PureLink HiPure Plasmid Miniprep Kit allows isolation of high yields of highly pure plasmid DNA from E.coli in less than 2 hours, without use of any organic solvents or cesium chloride, and contains Low endotoxin levels.

Plasmid DNA of High Purity

  • PureLink HiPure Plasmid Miniprep Kit is designed to efficiently isolate plasmid DNA from E.coli in 1.5–2 hours
  • Kit employs anion-exchange resin composed of small particles with a uniform pore size to purify plasmid DNA to a level equivalent to two passes through CsCl gradients
  • Need for additional steps to remove contaminants such as phenol, chloroform, and CsCl are eliminated, minimizing exposure to and disposal of hazardous materials
  • Efficient—Increased yields with low endotoxin Levels

High Yield and Efficient Endotoxin Removal

  • PureLink HiPure Plasmid Miniprep Kit allows purification of up to 30 μg of high quality plasmid DNA from 1–3 mL overnight E.coli cultures in approximately 1 hour, with an endotoxin level of 0.1–1 EU/μg, when cloning high copy number plasmids
  • Typically, DNA isolated using the PureLink HiPure Plasmid Miniprep Kit has an A260/A280 >1.80 when samples are diluted in Tris-HCl (pH 7.5) indicating that the DNA is reasonably free of proteins that could interfere with downstream applications

Isolated DNA can be used for a Number of Downstream Applications:

  • Purified DNA is suitable for downstream applications, including those requiring the highest purity DNA, such as mammalian transfections, automated and manual DNA sequencing, PCR amplification, in vitro transcription, bacterial cell transformation, cloning, and labeling

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 10 mL Resuspension Buffer (R3)
• 100 μL RNase A
• 10 mL Lysis Buffer (L7)
• 10 mL Precipitation Buffer (N3)
• 50 mL Equilibration Buffer (EQ1)
• 125 mL Wash Buffer (W8)
• 25 mL Elution Buffer (E4)
• 15 mL TE Buffer
• 25 HiPure Columns

Store all components at room temperature.
Format Kit
Isolation Technology Anion Exchange Resin
Scale Mini
Sample Type Bacterial Culture
Final Product Type BAC DNA, Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 25 Preps
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line PureLink
Product Type Plasmid MiniPrep Kit
Quantity 25 Preps
Shipping Condition Room Temperature
Target BAC DNA, Plasmid DNA
Test Time 2 hr.
Show More Show Less
compliance-icons
Product Identifier
  • PureLink HiPure Plasmid Miniprep Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulfate, sodium hydroxide

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.