missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ PCR Micro Kit

PureLink PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions.

Brand:  Invitrogen™ K310010

 View more versions of this product

Product Code. 10490344

  • £41.31 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: Columns, binding and wash buffers, recovery tubes, and the manual
This item is not returnable. View return policy

Description

Description

PureLink™ PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions.These highly purified DNA fragments and amplicons provide excellent results in downstream applications such as DNA sequencing, restriction digestion, cloning, PCR, and labeling. PureLink™ PCR Micro Kits provide a simple and rapid method for purifying and concentrating DNA fragments ranging in size from 100 bp-12 kb.

The unique micro-nozzle design allows up to 95% recovery of DNA fragments with as little as a 5μL elution volume. With less than a 10% coefficient of variance in the elution volume you can be sure that no ethanol is carried over from the wash steps giving you the best chance of success in downstream applications. Removal of primers, dNTPs, enzymes, and salts exceeds 98.5%. This kit can also efficiently remove unincorporated dye-labeled nucleotides from a cDNA labeling reaction.

Not High Throughput-Compatible (Manual)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature.
20 μL
Not High-throughput Compatible (Manual)
PCR Micro Kit
10 min.
Room Temperature
DNA from Enzymatic Reactions (e.g. PCR)
Silica Spin Column
DNA (Restriction Enzyme-Digested)
Spin Column
5 μL
PureLink™
10 Preps
100 bp to 12 kb
125 μL
Cloning, Standard PCR
10 Preps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.