Learn More
Invitrogen™ PureLink™ PCR Purification Binding Buffer B2
PureLink™ PCR Purification Binding Buffer B2 is the binding buffer used with the PureLink™ PCR Purification Kit.
Brand: Invitrogen™ K310005
35.66 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
PureLink™ PCR Purification Binding Buffer B2 is the binding buffer used with the PureLink™ PCR Purification Kit. It is added directly to PCR products in the first step of the PureLink™ PCR Purification Kit procedure to facilitate DNA binding to the silica membrane. The buffer is activated by the addition of 80mL of Isopropanol.
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. | |
PCR Purification Binding Buffer B2 | |
Room Temperature |
PureLink™ | |
120 mL |
For Research Use Only. Not for use in diagnostic procedures.