Learn More
Invitrogen™ PureLink™ PCR Purification Wash Buffer W1
The buffer is activated by the addition of 160 mL of 100% ethanol.
Brand: Invitrogen™ K310006
Code : 50
Additional Details : Weight : 0.50000kg
Description
The buffer is activated by the addition of 160 mL of 100% ethanol.
Not High Throughput-Compatible (Manual)
Order Info
Shipping Condition: Room Temperature
Specifications
Wash Buffer | |
PCR Products | |
Not High-throughput Compatible (Manual) | |
PCR Purification Wash Buffer W1 | |
Room Temperature |
Store the wash buffer at room temperature | |
DNA | |
PureLink™ | |
40 mL | |
PureLink™ |
For Research Use Only. Not for use in diagnostic procedures.