missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Quick Plasmid Miniprep Kit

Product Code. p-4926496
Click to view available options
No. of Reactions:
250 Preps
50 Preps
Unit Size:
50 preps
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10553293

Brand: Invitrogen™ K210010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Resuspension Buffer (R3)
RNase A (20 mg/mL)
Lysis Buffer (L7)
Precipitation Buffer (N4)
Wash Buffer (W9)
Wash Buffer (W10)
TE Buffer (TE)
Spin Columns
Wash and Recovery Tubes

The PureLink Quick Plasmid Miniprep Kit is designed for rapid isolation of sequencing-grade plasmid DNA from E.coli in about 30 minutes, using silica membrane columns.

The PureLink™ Quick Plasmid Miniprep Kit is designed for rapid isolation of sequencing-grade plasmid DNA from E.coli in 30-45 minutes, using silica membrane columns.

Features of the PureLink™ Quick Plasmid Miniprep Kit:

  • Fast—ready-to-use plasmid DNA typically in 30-45 minutes
  • Flexible—formats work with vacuum and centrifugation platforms

Reliable and cost-effective isolation of plasmid DNA

The PureLink™ Quick Plasmid Miniprep Kit allows isolation of high-quality plasmid DNA from 1–5 mL cultures in 30-45 minutes, using the PureLink™ spin columns. These columns use a unique silica membrane to achieve high yields of up to 40 μg of sequencing-grade plasmid DNA. Cells are lysed using an alkaline/SDS procedure and the lysate is applied to a silica membrane column that selectively binds plasmid DNA. Contaminants are removed with Wash Buffer, and the plasmid DNA is eluted in TE Buffer. The eluted DNA is suitable for all routine downstream applications.

Compatible with flexible formats

The PureLink™ Quick Plasmid Miniprep Kit can be used with a centrifuge or a vacuum manifold for purifying plasmid DNA. Typically, DNA isolated using the PureLink™ Quick Plasmid Miniprep Kit has an A260/A280 of >1.80 when samples are diluted in Tris-HCl (pH 7.5), indicating that the DNA is reasonably free of proteins that could interfere with downstream applications.

Isolated DNA can be used for a variety of downstream applications

Plasmid DNA purified using PureLink™ Quick Plasmid Miniprep Kit is suitable for all routine downstream applications, including bacterial and mammalian cell transformations, DNA sequencing, restriction enzyme digestion, cloning, and PCR.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 mg/mL
Column Type Spin Column
Content And Storage • Resuspension buffer
• RNase A (20 mg/mL)
• Lysis buffer
• Precipitation buffer
• Wash buffers
• TE buffer
• Spin columns in wash tubes
• Elution tubes

Format Spin Column
Isolation Technology Silica Spin Column
Sample Type Bacterial Culture
Final Product Type Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Cloning, Sequencing, Nucleic Acid Labeling, PCR, Transformation
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 50 Preps
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line PureLink
Product Type Quick Plasmid MiniPrep Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Target Plasmid DNA
Test Time 30 to 45 min.
Show More Show Less
compliance-icons
Product Identifier
  • PureLink Quick Plasmid Miniprep Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium azide, Sodiumhydroxide, Guanidine hydrochloride, Guanidine hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.