Learn More
Invitrogen™ pYES2/CT Yeast Expression Vector
S. cerevisiae expression vector derived from the parental pYES2 vector
Brand: Invitrogen™ V825120
Code : 50
Additional Details : Weight : 0.03000kg
Includes: Vectors are supplied supercoiled and lyophilized. Includes a lacZ expression control vector and an INVSc1 yeast stab.
Description
- Features the URA3 gene for selection in yeast
- pYES2/CT contains a C-terminal V5 epitope for detection and a polyhistidine (6xHis) tag for purification
Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Yeast Expression
Specifications
GAL1 | |
Vectors are supplied supercoiled and lyophilized. pYES2/NT is provided in three reading frames. Both pYES/NT and pYES/CT include a lacZ expression control vector and an INVSc1 yeast stab. Store vectors at -20°C. Store the stab at +4°C. All reagents are guaranteed stable for 6 months when properly stored. | |
His Tag (6x), V5 Epitope Tag | |
pYES |
Yeast Expression Vector | |
Ampicillin (AmpR) | |
Restriction Enzyme/MCS | |
YES™ |
For Research Use Only. Not for use in diagnostic procedures.