Learn More
Invitrogen™ pYES2/NT A, B, & C Yeast Expression Vectors
S. cerevisiae expression vector derived from the parental pYES2 vector
Brand: Invitrogen™ V825220
628.56 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.05000kg
Includes: Vectors are supplied supercoiled and lyophilized. pYES2/NT is provided in three reading frames. Includes a lacZ expression control vector and an INVSc1 yeast stab.
Description
- Offer the URA3 gene for selection in yeast
- pYES2/NT features an N-terminal Xpress™ epitope for detection with Anti-Xpress Antibody and a polyhistidine (6xHis) tag for purification with nickel-chelating resin
Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Yeast Expression
Specifications
GAL1 | |
Vectors are supplied supercoiled and lyophilized. pYES2/NT is provided in three reading frames. Both pYES/NT and pYES/CT include a lacZ expression control vector and an INVSc1 yeast stab. Store vectors at -20°C. Store the stab at +4°C. All reagents are guaranteed stable for 6 months when properly stored. | |
His Tag (6x), V5 Epitope Tag, Xpress Epitope Tag | |
pYES |
Yeast Expression Vector | |
Ampicillin (AmpR) | |
Restriction Enzyme/MCS | |
YES™ |
For Research Use Only. Not for use in diagnostic procedures.