Learn More
Invitrogen™ pYES2 Yeast Expression Vector
For native expression of the protein of interest in S. cerevisia
Brand: Invitrogen™ V82520
Code : 50
Additional Details : Weight : 0.06000kg
Includes: 20μg of expression vector, a TOP10F' E. coli stab, and an INVSc1 yeast stab.
Description
- Contains the URA3 gene for selection in yeast and 2μ origin for high-copy maintenance
Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Yeast Expression
Specifications
GAL1 | |
The pYES2 vector kit includes 20 μg of expression vector, a TOP10F´ E. coli stab, and an INVSc1 yeast stab. Store the vector at -20°C. Store the TOP10F´ and INVSc1 stabs at +4°C. All reagents are guaranteed stable for 6 months when stored properly. | |
Untagged | |
pYES |
Yeast Expression Vector | |
Ampicillin (AmpR) | |
Restriction Enzyme/MCS | |
YES™ |
For Research Use Only. Not for use in diagnostic procedures.