Learn More
Invitrogen™ pYES3/CT Yeast Expression Vector
S. cerevisiae expression vectors derived from the parental pYES2 vector
Brand: Invitrogen™ V825320
Code : 50
Additional Details : Weight : 0.04000kg
Includes: Vectors are supplied supercoiled and lyophilized and include a lacZ expression control vector and an INVSc1 yeast stab. The pYES6/CT Starter Kit also includes 50mg of Blasticidin.
Description
- Vector features a C-terminal V5 epitope for detection with an Anti-V5 Antibody
- pYES3/CT carries the TRP1 selection marker for selection in yeast
Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Yeast Expression
Specifications
Yeast Expression Vector | |
Ampicillin (AmpR) | |
Restriction Enzyme/MCS | |
YES™ |
Vectors are supplied supercoiled and lyophilized and include a lacZ expression control vector and an INVSc1 yeast stab. The pYES6/CT Starter Kit also includes 50 mg of Blasticidin. Store the vectors at -20°C. Store the INVSc1 stab and Blasticidin at +4°C. All reagents are guaranteed stable for 6 months when properly stored. |
|
His Tag (6x), V5 Epitope Tag | |
pYES |
For Research Use Only. Not for use in diagnostic procedures.