missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ Protein and Protein Broad Range (BR) Assay Kits

Achieve sensitive detection of protein, and low protein-to-protein variation, with the Qubit Protein and Protein BR Assay Kits for fluorescence-based protein quantitation assays.

Brand:  Invitrogen™ Q33212

 View more versions of this product

Product Code. 10788713

  • £295.00 / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Requires : Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes
This item is not returnable. View return policy

Description

Description

Quantify protein quickly and accurately with two unique Qubit protein quantification kits: the Qubit Protein Assay, which quantifies 12.5 μg/mL to 5 mg/mL of protein, and the Qubit Protein BR Assay Kit, which quantifies 0.1 to 20 mg/mL of protein. Both assays are compatible with many common contaminants, reducing reagents, salts, and nucleic acids. The Protein BR Assay Kit is also tolerant of detergents.

Qubit Protein Assay Kit
The Qubit Protein Assay Kit is designed specifically for use with any Qubit fluorometer. While using between 1 and 20 μL of sample, this assay can quantify samples ranging from 12.5 μg/mL to 5 mg/mL with low protein-to-protein variation.

The Qubit Protein Assay Kit is highly selective for proteins and is also highly accurate in the presence of reducing reagents, but not in the presence of large amounts of detergent. Common contaminants, such as reducing reagents (e.g., DTT, β-mercaptoethanol), salts, free nucleotides, amino acids, solvents, and DNA (but not detergents), are well tolerated in the assay. Slight protocol modifications are required for other contaminants.

The kit provides concentrated assay reagent, dilution buffer, and pre-diluted BSA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration.

Which product should you choose for fluorometric protein quantitation?
• 1–20 samples: use this Qubit Protein Assay Kit with the Qubit Fluorometer
• 20–2000 samples: use the Quant-iT Protein Assay Kit with a microplate reader

Qubit Protein BR Assay Kit
The Qubit Protein BR Assay Kit provides a quick, easy, and reliable method for determining the protein concentration of purified proteins, lysates, serum, plasma, or complex protein mixtures using a Qubit 4 Fluorometer. This protein kit can deliver accurate protein quantitation across a broad range (0.1–20 mg/mL), enabling most samples to be used neat (undiluted) and eliminating the guess work and dilution steps that accompany traditional protein quantitation methods. The assay is compatible with many common contaminants, such as detergents, reducing reagents (e.g., DTT, β-mercaptoethanol), salts, and nucleic acids.

Key features of the Qubit Protein BR Assay Kit include:
Broad dynamic range—0.1 to 20 mg/mL, measure samples without dilution
Two-point standard curve—eliminate time-consuming standard preparation
Automatic concentration determination—receive protein concentration instantaneously; no need for offline calculations
Compatible—accurate in the presence of reducing reagents, detergents, and other common buffer components

The Qubit Protein BR Assay Kit comes with two calibration standards, Protein BR Assay Buffer, and Protein BR Assay Reagent. Simply add 150–160 μL of Protein BR Assay Buffer to 10 or 20 μL of your sample, followed by 30 μL of Protein BR Assay Reagent. Incubate for 10 minutes at room temperature. Then insert the sample into a Qubit 4 instrument and read the concentration.

Notes
• The Qubit Protein Assay Kit can be used with Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers
• The Qubit Protein BR Assay Kit is a fluorescence-based assay intended for use only with the Qubit 4 Fluorometer
• 500 μL thin-walled clear PCR tubes (Cat. No. Q32856) are required but not included in the Protein Assay and BR Assay Kits

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Qubit Protein Assay
Qubit™
500 Reactions
12.5 μg/mL to 5 mg/mL
Room Temperature
Fluorescence
Protein Quantification Assay
Qubit Fluorometer, Microplate Reader
500 Assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit™ Protein and Protein Broad Range (BR) Assay Kits > 500 assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.