Learn More
Invitrogen™ RediLoad™ Loading Buffer
Designed for easy loading and tracking of PCR products run in agarose gels
Brand: Invitrogen™ 750026
118.56 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.13608kg
Description
RediLoad™ is added directly to the PCR reaction mixture prior to amplification. RediLoad features a reagent that increases density for easy loading, and a red tracking dye that migrates at approximately 25bp and allows easy monitoring of electrophoresis. RediLoad can be used as a replacement for conventional agarose loading buffer and is supplied in a 10X concentration. Compatible with agarose gels.
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store in refrigerator (2–8°C). | |
10X | |
Red dye (25 bp) | |
E-Gel™, Agarose Gels | |
Room Temperature |
Sample Loading Buffers | |
Agarose | |
Loading Buffer | |
Ready-Load™ |
For Research Use Only. Not for use in diagnostic procedures.