missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ribonuclease H

Product Code. 10368002
Click to view available options
Quantity:
120 U
30 U
Unit Size:
120 units
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10368002

Brand: Invitrogen™ 18021014

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Ribonuclease H containing 30 units supplied at a concentration of 2U/μL

Specifically degrades RNA strand of RNA-DNA hybrid to produce 5' phosphate-terminated oligoribonucleotides and single-stranded DNA

  • Source: Purified from E. coli expressing the E. coli RNase H gene on a plasmid
  • Performance and quality testing: Ribonuclease, nonspecific endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease
  • Unit definition: One unit hydrolyzes 1nmol of RNA in 3H-labeled poly(A)poly(dT) to acid-soluble material in 20 min. at 37°C
  • Unit reaction conditions: 20mM Tris-HCl (pH 7.5), 0.1M KCl, 10mM MgCl2 , 0.1mM DTT, 5% (w/v) sucrose, 0.5nmol 3H-labeled poly(A)poly(dT), and enzyme in 50μL for 20 min. at 37°C

Removal of mRNA during second-strand cDNA synthesis; Removal of poly(A) sequences from mRNA in the presence of oligo(dT); Oligodeoxyribonucleotide-directed cleavage of RNA; PCR & Real-Time PCR; Reverse Transcription

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage 1 vial of Ribonuclease H containing 30 units supplied at a concentration of 2U/μl

Store at -20°C in a non-frost-free freezer.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme RNase
Quantity 30 U
Product Type Ribonuclease H

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.