Learn More
Invitrogen™ Ribonuclease Inhibitor, Cloned
Neutral pancreatic ribonuclease inhibitor
Brand: Invitrogen™ 15518012
163.40 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02000kg
Includes: Ribonuclease Inhibitor, Cloned is supplied at 10,000 units/mL in 20mM HEPES-KOH (pH 7.6), 50mM KCl, 8mM DTT, and 50% (v/v) glycerol.
Description
Ribonuclease Inhibitor, Cloned is a potent non-competitive inhibitor of neutral pancreatic ribonucleases.
Applications: Cell-free translation systems. Reverse transcription of mRNA.
Source: Recombinant, purified from E. coli.
Performance and Quality Testing: No detectable contaminating activity is observed in DNA nicking, ribonuclease, and protease assays.
Unit Definition: One unit inhibits 5ng RNase A by 50% using cytidine 2', 3' cyclic monophosphate (cCMP) as the substrate.
PCR and Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Dry ice
Specifications
50 mM | |
Ribonuclease Inhibitor, Cloned is supplied at 10,000 units/ml in 20 mM HEPES-KOH (pH 7.6), 50 mM KCl, 8 mM DTT, and 50% (v/v) glycerol. Store at -20°C. | |
1000 U | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.