Learn More
Invitrogen™ S.N.A.P.™ UV-Free Gel Purification Kit
The S.N.A.P.™ UV-Free Gel Purification Kit uses non-toxic reagents to rapidly purify intact DNA fragments from agarose gels.
Brand: Invitrogen™ K200025
Code : 50
Additional Details : Weight : 0.56000kg
Includes: Each S.N.A.P. UV-Free Gel Purification Kit includes enough crystal violet reagents, S.N.A.P. columns, collection vials, sodium iodide solution, purification buffers, and sterile water for 25 gel purifications.
Description
The S.N.A.P.™ UV-Free Gel Purification Kit uses non-toxic reagents to rapidly purify intact DNA fragments from agarose gels. The key to isolating undamaged DNA is the unique crystal violet reagent, which allows visualization of DNA (≥200 ng) in the gel under ambient light. This eliminates the need for ethidium bromide and exposure to UV light, which can nick and damage DNA. Nucleic acids up to 7 kb are efficiently purified from the gel using the convenient S.N.A.P.™ resin, provided ready to use in a spin column format. The resin eliminates organic extractions, which generate hazardous waste. In addition, there are no time-consuming ethanol precipitations.
Easy to Use
The S.N.A.P.™ UV-Free Gel Purification Kit yields pure, intact DNA in an easy-to-perform, rapid procedure. Simply:
1. Prepare gel with crystal violet reagent and load samples in crystal violet loading buffer.
2. Visualize DNA band under ambient light and excise from agarose gel.
3. Add NaI solution (provided) and melt the agarose at 42-50°C.
4. Load sample onto the S.N.A.P.™ spin column.
5. Wash away contaminants.
6. Elute DNA in water.
The eluted DNA is highly pure, intact, and ready to use for cloning, sequencing, transformation, or transfection.
Specifications
Spin Column | |
Kit | |
DNA Fragments in Agarose Gel Slices, Gel Samples | |
Not High-throughput Compatible (Manual) | |
25 Preps | |
Gel Purification Kit | |
Nucleic Acids in Gel Fragments |
Each S.N.A.P.™ UV-Free Gel Purification Kit includes enough crystal violet reagents, S.N.A.P.™ columns, collection vials, sodium iodide solution, purification buffers, and sterile water for 25 gel purifications. Store all components at room temperature. All reagents are guaranteed stable for 6 months when properly stored. | |
Spin Column | |
Cloning, PCR | |
Crystal Violet | |
S.N.A.P. ™ | |
25 Preps |
For Research Use Only. Not for use in diagnostic procedures.