Learn More
Invitrogen™ SuperScript™ Double-Stranded cDNA Synthesis Kit
Maximize the yield of full-length cDNA, as well as that of overall cDNA synthesis
£1578.00 - £6990.00
Specifications
Format | Kit |
---|---|
Labeling Method | Direct Labeling |
Optimal Reaction Temperature | 42°C |
Product Line | SuperScript™ |
Product Type | cDNA Synthesis Kit |
Includes: Reagents and buffers for 10 first- and second-strand cDNA synthesis reactions converting 25-50μg total RNA or 0.2-5μg mRNA into double-stranded cDNA.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10729803
|
Invitrogen™
11917010 |
10 Reactions |
£1578.00
10 reactions |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073372
|
Invitrogen™
11917020 |
50 Reactions |
£6990.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Contains all of the reagents (except an oligo (dT)-containing primer) necessary to make high-quality, double-stranded cDNA from total RNA or poly A+-selected RNA (mRNA).
Simple, reliable, and trouble-free
The SuperScript Double-Stranded cDNA Synthesis Kit offers a simple, reliable, and trouble-free method by which RNA templates can be converted to cDNA for use in cloning and library construction experiments. The reactions described in the SuperScript Double-Stranded cDNA Synthesis Kit protocol are designed to convert 25–50μg total RNA, or 0.2–5μg mRNA, into first- and second-strand cDNA (supply your own primer). Enjoy the benefits of a tested protocol and reagents of consistent quality. To get the best possible cDNA product using this kit, use a high-quality RNA substrate.
- Reliability: each buffer, reagent, and enzyme in the kit is of the same high quality you expect from Life Technologies
- Performance: SuperScript II is an enzyme you can trust for high-yield, high-quality, general-purpose, first-strand synthesis
- Optimal temperature: 42°C
DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis, PCR and Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Wet Ice
Specifications
Kit | |
42°C | |
cDNA Synthesis Kit | |
Wet Ice | |
cDNA (Double-Stranded) |
Direct Labeling | |
SuperScript™ | |
SuperScript™ II | |
Microarray Analysis | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.