Learn More
Invitrogen™ SuperScript™ II Reverse Transcriptase
Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT.
Brand: Invitrogen™ 18064014
255.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT. Mutations in the RNase H domain of the enzyme eliminate degradation of the RNA during first-strand cDNA synthesis, which results in higher yields of full-length cDNA.
SuperScript reverse transcriptases are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.
Features of SuperScript II Reverse Transcriptase include:
• High cDNA yields and more full-length product
• Generation of cDNA up to 12.3 kb
• First-strand cDNA synthesis at higher temperatures than conventional MMLV RT
Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase (Cat. No. 18090010), features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.
Order Info
Shipping Condition: Wet ice

Specifications
• SuperScript II Reverse Transcriptase, 50 μL (10,000 units at 200 U/μL) |
|
High | |
Reverse Transcription | |
10,000 Units | |
Reduced | |
200 U/μL | |
50 Reactions | |
Reverse Transcription | |
RNA |
Stand-alone Enzyme | |
50 min. | |
42°C | |
SuperScript II | |
Wet Ice | |
First-Strand cDNA | |
Separate components | |
Up to 12.3 kb |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3037372 | Certificate of Analysis | 31/01/2025 | 18064014 |
3074081 | Certificate of Analysis | 31/01/2025 | 18064014 |
2467960 | Certificate of Analysis | 29/01/2025 | 18064014 |
2515632 | Certificate of Analysis | 29/01/2025 | 18064014 |
2487082 | Certificate of Analysis | 29/01/2025 | 18064014 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.