Learn More
Invitrogen™ SuperScript™ III Reverse Transcriptase
Invitrogen SuperScript III Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) created by the introduction of several mutations for reduced RNase H activity, increased half-life, and improved thermal stability.
Brand: Invitrogen™ 18080093
59.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Invitrogen SuperScript III Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) created by the introduction of several mutations for reduced RNase H activity, increased half-life, and improved thermal stability. SuperScript III RT offers higher cDNA yields, improved cDNA lengths, improved efficiency on GC-rich target RNAs, and overall better performance than wild-type MMLV and MMLV RNase H-minus enzymes.
SuperScript RTs are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.
Features of SuperScript III Reverse Transcriptase include:
• First-strand cDNA may be synthesized at temperatures up to 55°C
• cDNA can be generated from 100 bp to >12 kb
• Increased specificity
• High yields of cDNA and full-length products
Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase (Cat. No. 18090010), features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.
Order Info
Shipping Condition: Dry ice

Specifications
• SuperScript™ III Reverse Transcriptase, 10 μL (2,000 units at 200 U/μL) |
|
High | |
Reverse Transcription | |
2,000 Units | |
Reduced | |
200 U/μL | |
10 Reactions | |
Reverse Transcription | |
RNA |
Stand-alone Enzyme | |
30 to 50 min. | |
50°C | |
SuperScript III | |
Dry Ice | |
First-Strand cDNA | |
Separate components | |
Up to 12.3 kb |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3041351 | Certificate of Analysis | 31/01/2025 | 18080093 |
2489772 | Certificate of Analysis | 29/01/2025 | 18080093 |
2539873 | Certificate of Analysis | 29/01/2025 | 18080093 |
2614615 | Certificate of Analysis | 29/01/2025 | 18080093 |
2735897 | Certificate of Analysis | 29/01/2025 | 18080093 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.