missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ VILO™ cDNA Synthesis Kit

The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes.

Brand:  Invitrogen™ 11754250

1715.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10704274

  • £2450.00 / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: 5X VILO Reaction Mix (200μL) and a 10X SuperScript III Enzyme Blend (100μL), which contains: SuperScript III RT, RNaseOUT Recombinant Ribonuclease Inhibitor, and a proprietary helper protein
This item is not returnable. View return policy

Description

Description

The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes. The enzyme blend contains SuperScript III Reverse Transcriptase (RT), a genetically engineered MMLV RT that has reduced RNase H activity and improved thermostability for highly efficient cDNA synthesis.

The SuperScript VILO cDNA Synthesis Kit can be used to synthesize cDNA from for a wide range of input RNA amounts. This kit provides the high temperature capability of SuperScript III Reverse Transcriptase in an optimized format for generating first-strand cDNA for use in RT–qPCR.

Notes
• For superior first-strand cDNA synthesis in two-step RT-qPCR, try SuperScript IV VILO Master Mix (Cat. No. 11756050) or SuperScript IV VILO Master Mix with ezDNase Enzyme (Cat. No. 11766050).
• This kit is included with EXPRESS Two-Step RT–qPCR kits. Reagents are provided for 50 or 250 20-μL cDNA synthesis reactions.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 5X VILO Reaction Mix, 1,000 μL
• 10X SuperScript III Enzyme Blend, 500 μL

Store at –5 to –30°C.

Kit
60 min.
42°C
Reduced
Real Time PCR (qPCR)
250 Reactions
Reverse Transcription
RNA
qPCR
High
Reverse Transcription
SuperScript III
Dry Ice
First-Strand cDNA
Master Mix
Up to 10 kb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.