missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO

Extremely sensitive stain for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels

Brand:  Invitrogen™ S7567

450.80 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10207252

  • £644.00 / 1mL
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

SYBR Green I offers greater sensitivity for dsDNA, making it especially useful for assays where the presence of contaminating RNA or ssDNA might obscure results. With exceptionally low background fluorescence and spectral characteristics that closely match light sources and filter sets in existing instruments, SYBR Green I stain is ideal for use with laser scanners.
Product use 1mL stains 100 minigels.

  • Sensitivity: at least four times greater than ethidium bromide for DNA in agarose gels
  • Ease of use: gels soaked in diluted stain can be visualized without desalting
  • Compatibility: with UV transilluminators, gel documentation systems, and laser scanners
  • Flexibility: for use with a broad range of applications, including DNA typing, PCR-based assays, DNA damage assays, analysis of complex samples, and real-time PCR detection

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Fluorescence
Nucleic Acid Gel Stain
Room Temperature
In-Gel Detection
SYBR Green I
1 mL
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.