Learn More
Invitrogen™ SYBR™ Green Nucleic Acid Gel Stain Starter Kit
Everything needed for first time users to use SYBR gel stains
Brand: Invitrogen™ S7580
163.40 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Includes: Kit contains: 50μL of SYBR Green I nucleic acid gel stain (S-7567), 50μL of SYBR Green II RNA gel stain (S-7568) and a SYBR Green/Gold gel stain photographic filter (S-7569).
Description
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
In-Gel Detection | |
SYBR Green II, SYBR Green I | |
RNA, DNA |
Fluorescence | |
Nucleic Acid Gel Stain Starter Kit |
For Research Use Only. Not for use in diagnostic procedures.