missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Safe DNA Gel Stain in 0.5X TBE
GREENER_CHOICE

Product Code. p-4926801
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10534623

Brand: Invitrogen™ S33100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Developed specifically for reduced mutagenicity – to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels

SYBR Safe stain is less mutagenic than ethidium bromide and offers better sensitivity than ethidium bromide. Supplied as a premixed solution that can be used like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe stain has fluorescence excitation maxima at ∽280 and ∽502nm, and an emission maximum at ∽530nm. SYBR Safe DNA gel stain is also available as 4L (S-33101) of ready-to-use solution. The SYBR Safe DNA Gel Stain Starter Kit (S-33110, sold separately) includes 1L of SYBR Safe DNA gel stain and one photographic filter (S-37100).

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Supplied as a ready-to-use solution in 0.5X TBE

Store at room temperature in the original container.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYBR Safe
Product Type DNA Gel Stain
Green Features Less hazardous
Quantity 1 L
Shipping Condition Room Temperature
Target Molecule DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.