missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE
GREENER_CHOICE

Product Code. p-4926802
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11560676

Brand: Invitrogen™ S33112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Developed specifically for reduced mutagenicity to be less hazardous. Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE is safer and more sensitive than ethidium bromide for staining DNA in agarose or acrylamide gels. The premixed solution is available in 1L and 4L units.

SYBR Safe™ stain is not only less mutagenic than ethidium bromide, but SYBR Safe™ stain′s detection sensitivity is better than that of ethidium bromide. SYBR Safe™ stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe™ stain has fluorescence excitation maxima at ∼280 and ∼502nm, and an emission maximum at ∼530nm.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Supplied as a ready-to-use solution in 1X TAE

Store at room temperature in the original container.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYBR Safe
Product Type DNA Gel Stain
Green Features Less hazardous
Quantity 4 L
Shipping Condition Room Temperature
Target Molecule DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.