Learn More
Invitrogen™ T-REx™-293 Cell Line
Significant time and effort savings when using the T-REx™ System
Brand: Invitrogen™ R71007
Code : 50
Additional Details : Weight : 0.00525kg
Description
T-REx Cell Lines stably express the tetracycline repressor protein. The T-REx Cell Lines are functionally tested by transient transfection with the positive control vector pcDNA™4/TO/lacZ. T-REx Cell Lines exhibit extremely low basal expression levels in the repressed state and high expression upon induction with tetracycline.
Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression
Specifications
1 x 107 cells are supplied frozen in 1 ml 90% complete medium and 10% DMSO. Store in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored. | |
Human | |
3 x 10^6 Cell |
T-REx-293 Cell Line | |
T-REx™ |
For Research Use Only. Not for use in diagnostic procedures.