Learn More
Invitrogen™ T-REx™-HeLa Cell Line
Product Code. 10032662
Click to view available options
Quantity:
1 x 107 cells
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 10032662
Brand: Invitrogen™ R71407
For Research Use Only. All usage must comply with product instructions.
Stably express the tetracycline repressor protein
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 x 107 cells are supplied frozen in 1 ml 90% complete medium and 10% DMSO. Store in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored.
Cell Line T-REx-HeLa Cell Line
Species Human
No. of Cells 1 x 107
Product Line T-REx
Quantity 1 x 107 cells