missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ T4 DNA Polymerase
DNA polymerase with 3'-exodeoxyribonuclease activity, but not 5'a3' exodeoxyribonuclease activity
£162.00 - £548.00
Specifications
Content And Storage | T4 DNA Polymerase is supplied with a vial of 5X T4 DNA Polymerase buffer [165 mM Tris acetate (pH 7.9), 330 mM sodium acetate, 50 mM magnesium acetate, 5 mM DTT]. Store at -20°C. |
---|---|
Description | DNA Polymerase |
Polymerase | T4 DNA Polymerase |
Exonuclease Activity | 3' - 5' |
Shipping Condition | Wet Ice |
Includes: T4 DNA Polymerase is supplied with a vial of 5X T4 DNA Polymerase buffer [165mM Tris acetate (pH 7.9), 330mM sodium acetate, 50mM magnesium acetate, 5mM DTT].
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10164552
|
Invitrogen™
18005017 |
50 U |
£162.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10043202
|
Invitrogen™
18005025 |
250 U |
£548.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Applications: Labeling double-stranded linear DNA by replacement synthesis, Oligonucleotide-directed, site-specific mutagenesis, 3' end-labeling of double-stranded DNA, Polishing both 5' or 3 overhangs to make blunt ends
- Source: Purified from E. coli expressing the T4 DNA Polymerase gene on a plasmid
- Performance and Quality Testing: Single- and double-stranded endodeoxyribonuclease and phosphatase assays; exodeoxyribonuclease and polymerase activities tested
- Unit Definition: One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using DNase I-nicked DNA as template primer
- Unit Reaction Conditions: 50mM glycine-NaOH (pH 8.8), 16.6mM (NH4)2SO4, 6mM MgCl2, 6.5μM EDTA, 10mM 2-mercaptoethanol, 0.165mg/mL BSA, 1.6mg/mL DNase I-nicked salmon testes DNA, 0.33mM dCTP, 0.33mM dATP, 0.33mM dGTP, 0.33mM dTTP, 76nM [3H]dTTP, and enzyme in 0.1mL for 30 min. at 37°C
- A T4 DNA Polymerase Technical Bulletin is available
ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics and Non-Coding RNA Research, Restriction Enzyme Cloning
Order Info
Shipping Condition: Wet Ice
Specifications
T4 DNA Polymerase is supplied with a vial of 5X T4 DNA Polymerase buffer [165 mM Tris acetate (pH 7.9), 330 mM sodium acetate, 50 mM magnesium acetate, 5 mM DTT]. Store at -20°C. | |
T4 DNA Polymerase | |
Wet Ice |
DNA Polymerase | |
3' - 5' | |
No |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ T4 DNA Polymerase