Learn More
Invitrogen™ T7 RNA Polymerase
DNA-dependent RNA polymerase with high specificity for bacteriophage T7 promoter sequences
£221.00 - £392.00
Specifications
Polymerase | T7 RNA Polymerase |
---|---|
Promoter | T7 |
Includes: T7 RNA Polymerase is supplied with a vial of 5X reaction buffer [200mM Tris-HCl (pH 8.0), 40mM MgCl2 , 10mM spermidine-(HCl)3 , 125mM NaCl], vial of 100mM DTT.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10093062
|
Invitrogen™
18033019 |
2500 U |
£221.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11528906
|
Invitrogen™
18033100 |
2 x 2500 U |
£392.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synthesizes large quantities of RNA from DNA inserted into a transcription vector downstream from a T7 promoter. A T7 RNA Polymerase technical bulletin is available.
- Application: Synthesis of labeled and unlabeled RNA transcripts.
- Source: Purified from E. coli expressing the T7 RNA Polymerase gene on a plasmid
- Performance and Quality Testing: 3' and 5' exodeoxyribonuclease, ribonuclease, and DNA nicking assays; performance in a transcription reaction
- Unit Definition: One unit hydrolyzes 1nMol of ribonucleotide into acid-precipitable material in 1 hr at 37°C using a T7 transcription vector as template
- Unit Reaction Conditions: 40mM Tris-HCl (pH 8.0), 25mM NaCl, 8mM MgCl2, 2mM spermidine-(HCl)3, 5mM DTT, 0.4mM ATP, 0.4mM CTP, 0.4mM GTP, 0.4mM UTP, 1μCi [3H]GTP, 1μg SphI-cut pT7L13, and enzyme in 50μL for 10 min. at 37°C
Cell Free Protein Expression for Functional Analysis, Cell Free Protein Expression for Structural Analysis, Cell-Free (in vitro) Expression, Gene Expression Analysis and Genotyping, In Vitro Transcription, Protein Expression, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
T7 RNA Polymerase | |
T7 |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.