missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ INVαF' Chemically Competent E. coli
Quick, one-step cloning strategy for directly inserting Taq-amplified PCR product into plasmid vector
£441.00 - £714.00
Specifications
Bacterial or Yeast Strain | INVαF ́ |
---|---|
Cell Type | Chemically Competent E. coli |
Cloning Method | TA Cloning |
Content And Storage | TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
Format | Kit |
Product Code | Brand | No. of Reactions | Vector | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Vector | Price | Quantity & Availability | ||||
10646533
|
Invitrogen™
K200001 |
20 Reactions | pCRII |
£441.00
Each |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10573263
|
Invitrogen™
K200040 |
40 Reactions | pCR2.1 |
£714.00
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The TA Cloning Kit uses the pCR2.1 cloning vector and ExpressLink T4 DNA Ligase to generate a ligation product in a 15-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.
- Fast and convenient: 15-minute, room-temperature ligation
- Efficient: blue/white screening and >80% clones with correct insert
- Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
- Hassle-free: eliminates any enzymatic modifications of PCR product
- Streamlined: does not require use of PCR primers that contain restriction sites
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- T7 promoter for in vitro RNA transcription and sequencing
- Versatile polylinker with flanking EcoR I sites for easy excision of inserts
- M13 forward and reverse primer sites for sequencing
- Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to the 3' ends of PCR products
- Linearized vector supplied in has single 3' deoxythymidine (T) residues
- Allows PCR inserts to ligate efficiently with vector
Cloning, PCR Cloning
Specifications
INVαF ́ | |
TA Cloning | |
Kit | |
One Shot™ | |
T7 |
Chemically Competent E. coli | |
TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
|
PCR Cloning | |
Cloning Kit |
For Research Use Only. Not for use in diagnostic procedures.