missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli

Provides a quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into a plasmid vector. Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli is available in 20- and 40-reaction kit sizes.

Brand:  Invitrogen™ K204001

424.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : 50

Additional Details : Weight : 0.00500kg

 View more versions of this product

Product Code. 10093092

  • £447.00 / Each
In Stock
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

The TA Cloning™ Kit uses the pCR™2.1 cloning vector and ExpressLink™ T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

  • Fast & Convenient—15-minute, room-temperature ligation
  • Efficient—Blue/white screening and >80% clones with correct insert
  • Flexible—Choice of kanamycin or ampicillin resistance for flexible antibiotic selection
  • Hassle-free—Eliminates any enzymatic modifications of the PCR product
  • Streamlined—Does not require the use of PCR primers that contain restriction sites

The pCR™2.1 vector provides:

  • 3′-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 promoter for in vitro RNA transcription and sequencing
  • Versatile polylinker with flanking EcoR I sites for easy excision of inserts
  • M13 forward and reverse primer sites for sequencing

How TA Cloning™ Works
Taq polymerase has a non-template-dependent activity that adds a single deoxyadenosine (A) to the 3′ ends of PCR products. The linearized vector supplied in this kit has single 3′ deoxythymidine (T) residues. This allows PCR inserts to ligate efficiently with the vector.

Other Kit Configurations
TA Cloning™ Kits are also offered without competent cells (K2020-20, K2020-40), with One Shot™ INVF Chemically Competent E. coli (K2000-01, K2000-40) or with One Shot™ TOP10F Chemically Competent E. coli (K2030-01, K2030-40).

Specifications

Specifications

TOP10
TA Cloning
Kit
20 Reactions
Cloning Kit
pCRII
Chemically Competent E. coli
TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
PCR Cloning
One Shot™
T7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.