Learn More
Invitrogen™ TE Buffer (20X), RNase-free
Essential to success of RiboGreen™ RNA quantitation assays
Brand: Invitrogen™ T11493
151.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
RNase-free TE Buffer (20X) is an essential component of Qubit and Quant-iT assays, including PicoGreen, RiboGreen, and Qubit dsDNA HS. It is offered here as a standalone solution to support quantification of dsDNA, ssDNA and RNA.
Note: For dilution instructions, storage considerations, and assay compatibility information please see the Product Information Sheet (Pub. No. MAN1000402) below.
Order Info
Shipping Condition: Room temperature

Specifications
Store at room temperature. | |
TE Buffer | |
Room Temperature |
20X | |
100 mL | |
Liquid |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.