missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tissue Extraction Reagent II

Lysis buffer for total protein extraction from tissue samples

Brand:  Invitrogen™ FNN0081

Product Code. 10101203

  • £121.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Build Your Own Immunoassay, Cell Analysis, Cell Lysis, Immunoassays, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
Luminex Assay, Western Blot, Sample preparation for ELISA
Dry Ice
Tissue Extraction Reagent
100 ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.