Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit for Subcloning, with TOP10 E. coli, without manual
Brand: Invitrogen™ KNM450040
This item is not returnable.
View return policy
Description
Specifications
Kit, Cloning for Subcloning | |
TOP10 | |
50 Reactions |
Box 1: Topoisomerase I-activated pCR2.1-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid | |
Cloning, PCR Cloning |