Learn More
Invitrogen™ Total Exosome Isolation Reagent (from other body fluids)
Total Exosome Isolation Reagent from other body fluids enables fast and efficient enrichment of intact exosomes from cerebrospinal fluid (CSF), ascitic fluid, amniotic fluid, milk, and saliva with a protocol that is scalable depending on your sample size.
Brand: Invitrogen™ 4484453
277.10 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Total Exosome Isolation Reagent from other body fluids enables fast and efficient enrichment of intact exosomes from cerebrospinal fluid (CSF), ascitic fluid, amniotic fluid, milk, and saliva with a protocol that is scalable depending on your sample size.
- Maximize recovery of intact exosomes for any type of downstream application
- Easily isolate exosomes using our simple and reliable protocol
- Avoid time-consuming ultra-centrifugation
- Maximize flexibility—can be scaled up or down depending on the sample size

Specifications
Store at 2–8°C. | |
Exosomes | |
6 mL |
Total Exosome Isolation | |
Liquid |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2798710 | Certificate of Analysis | 29/01/2025 | 4484453 |
2823278 | Certificate of Analysis | 29/01/2025 | 4484453 |
2809239 | Certificate of Analysis | 29/01/2025 | 4484453 |
2835115 | Certificate of Analysis | 29/01/2025 | 4484453 |
2853079 | Certificate of Analysis | 28/01/2025 | 4484453 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.