missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Total RNA Lysis Solution, Nucleic Acid Purification

Total RNA Lysis Solution is a 2X concentrate ideal for the chemical disruption of cells and tissue (cultured cells, primary cell isolates, and animal/plant tissues) prior to RNA or DNA extraction.

Brand:  Invitrogen™ 4305895

111.75 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15154328

  • £149.00 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Total RNA Lysis Solution is a 2X concentrate ideal for the chemical disruption of cells and tissue (cultured cells, primary cell isolates, and animal/plant tissues) prior to RNA or DNA extraction. Following downstream purification (e.g., using the 6100 PrepStation), intact RNA can be isolated that is free of contaminating protein and metal ions. Total RNA Lysis solution is the recommended reagent for the Total RNA Isolation Chemistry System for the ABI PRISM™ 6100 Nucleic Acid PrepStation.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2X
6100 Nucleic Acid PrepStation
250 mL
Store at room temperature.
100 Preps
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.