missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrackIt™ 100 bp DNA Ladder
GREENER_CHOICE

Product Code. 10778594
Click to view available options
Quantity:
50 μg
Unit Size:
100 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10778594

Brand: Invitrogen™ 10488058

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Contains: TrackIt™ 100bp DNA Ladder (0.1μg/μL) in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.6% tartrazine, and 5% glycerol; For 100 applications

Invitrogen TrackIt 100 bp DNA Ladder is ready to use and designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp.

Invitrogen TrackIt 100 bp DNA Ladder is ready to use and designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. 100 bp DNA Ladder consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 1500, and 600 bp for easy orientation.

TrackIt 100 bp DNA Ladder is ideal for separation on 1–2% agarose gels.

Highlights of TrackIt 100 bp DNA Ladder:

  • Sharp, clear bands—chromatography purified fragments for consistent and reliable results and stable at room temperature up to six months
  • Ready-to-use—premixed with 6X TrackIt Cyan/Orange Loading Buffer for tracking of DNA migration
  • Precise—an exact amount of DNA in each band

Product use

The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 500 μL TrackIt 100 bp DNA Ladder
• 1 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at room temperature or at 4°C for up to 24 months. For longer periods, store at -20°C.
Concentration 0.1 μg/μL
Gel Type Agarose
Product Type DNA Ladder
Ready to Load Yes
Sample Loading Volume 1 mL
Size Range 100 to 2000 bp
Volume (Metric) 50 μg
Gel Compatibility Agarose gel
Green Features Sustainable packaging
No. of Reactions 100 Applications
Product Line TrackIt
Quantity 50 μg
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.