Learn More
Invitrogen™ TrackIt™ 25 bp DNA Ladder
Combines DNA Ladder with loading buffer in a single tube
Brand: Invitrogen™ 10488022
Code : 50
Additional Details : Weight : 0.02000kg
Description
The TrackIt™ format minimizes time spent thawing, diluting, and adding sample buffer to DNA ladders. Simply transfer your TrackIt DNA Ladder from the vial to the gel. These ladders are stable at room temperature and can be stored on the bench top. TrackIt DNA Ladders offer two tracking dyes, one that runs behind the sample and one that runs ahead of the sample. Dyes serve as visual markers for following migration during electrophoresis and indicate when maximum resolution of the DNA fragments has been achieved. Additionally, visualization of DNA bands will not be obscured by the tracking dyes because they run outside the limits of most DNA samples.
- Ready-to-load convenience
- Better tracking with two dyes to indicate when maximum resolution has been achieved
- TrackIt dyes that run outside the limits of most DNA samples, minimizing masking of sample DNA bands
- Compatible with agarose andE-Gel™ gels
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice
Specifications
0.5 μg/μL | |
Liquid | |
20 Applications |
DNA Molecular Weight Marker/Ladder | |
TrackIt™ 10bp DNA Ladder is supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.4% Orange G, and 5% glycerol; TrackIt™ 25bp DNA Ladder is supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.4% Orange G, and 5% glycerol; TrackIt™ 50bp DNA Ladder is supplied at 0.1μL/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.4% Orange G, and 5% glycerol; TrackIt™ 100bp DNA Ladder is supplied at 0.1μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.6% tartrazine, and 5% glycerol; TrackIt™ 1Kb Plus DNA Ladder is supplied at 0.1μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.6% tartrazine, and 5% glycerol; TrackIt™ 1 Kb DNA Ladder is supplied at 0.1μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.6% tartrazine, and 5% glycerol; TrackIt™ DNA/Hind III Fragments are supplied at 0.1μg/μL in 10mM | |
25 to 500 bp |
For Research Use Only. Not for use in diagnostic procedures.